


1-Butanol ≥99.0%, Ultrapure for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:14,1540
Boiling Pt:117…118 °C
Melting Pt:–90 °C
Density:0.810
Flash Pt:35 °C (95 °F)
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS05
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Related Information
Frequently Bought Together