


1-Butanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (760 mmHg)
Melting Pt:–89 °C
Density:0.81 g/cm³
Flash Pt:35 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS07
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3980 to 1.4000 (20°C, 589 nm)
- Water:≤0.2 % (Coulometric)
- GC:≥98.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specification Test Results
Frequently Bought Together