

Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:1155
ADR:3,I
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 4-7 ppm
EINECS:200-467-2
REACH:01-2119535785-29
:GHS02
:GHS07
Specification Test Results
- Assay:Min. 99 %
- Assay (Ionol/BHT):4 - 10 ppm
- Acidity:Max. 0.0003 meq/g
- Evaporation residue:Max. 20 ppm
- Water:Max. 0.2 %
Specification Test Results
Frequently Bought Together