Heptylamine 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-68-2
:
Formula:H₃C(CH₂)₅CH₂NH₂
MDL Number:MFCD00008244
:115.22 g/mol
Boiling Pt:153…156 °C (760 mmHg)
Melting Pt:–23 °C
Density:0.774 g/cm³
Flash Pt:44 °C
Storage Temperature:Ambient
EINECS:203-895-8
UN:2734
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4233 to 1.4253 (20°C, 589 nm)
- Water:≤0.3 %
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together