Order Entry
Austria
ContactUsLinkComponent
Triethylamine for synthesis, Sigma-Aldrich®
Triethylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Triethylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    133 g/l (20 °C)
  • Melting Point:
    -115 °C
  • Molar Mass:
    101.19 g/mol
  • Boiling Point:
    90 °C (1013 hPa)
  • Vapor Pressure:
    69 hPa (20 °C)
  • Flash Point:
    -11 °C
  • Explosion Limit:
    1.2 - 9.3 %(V)
  • Density:
    0.73 g/cm3 (20 °C)
  • Ignition Point:
    215 °C
  • Saturation concentration:
    (20 °C) Air,no data available
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Frequently Bought Together