Trifluoroacetic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: TFA
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:72 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
UN:2699
ADR:8,I
:M-Clarity™ quality level = MQ200
EINECS:200-929-3
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-15 °C
- Molar Mass:114.02 g/mol
- Boiling Point:72 °C (1013 hPa)
- Vapor Pressure:141 hPa (20 °C)
- Density:1.48 g/cm3 (20 °C)
- pH:1 (10 g/l, H2O)
Specification Test Results
Related Information
Frequently Bought Together