


Trichloroacetic acid (TCA) ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS05
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 98 %
- Melting point:55 - 59 °C
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.1 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
spec_VWRC20734.295 spec_VWRC20734.460 spec_VWRC20734.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together