Order Entry
Austria
ContactUsLinkComponent
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Health
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier:  VWR Chemicals
Certificates
undefined
Health
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 10 ppm

Specification Test Results

Related Information

Frequently Bought Together