Order Entry
Austria
ContactUsLinkComponent
 
 
Dichloromethane, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised, AnalaR NORMAPUR® analytical reagent
  25630.290
 :  
undefined
 
Dichloromethane, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised, AnalaR NORMAPUR® analytical reagent
  25630.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    Clear colourless liquid
  • Acidity:
    Max. 0.0010 %
  • Colour value:
    Max. 10 APHA
  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Water (K.F.):
    Max. 0.0050 %
  • Chloromethane:
    Max. 0.0100 %
  • Carbon tetrachloride:
    Max. 0.0100 %
  • Substances discoloured by H2SO4:
    Max. 60 APHA
  • Free chlorine:
    Max. 0.00003 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.3 ppm

 

 

 :
 :84.93 g/mol
 :12,06140
 :39.8 °C (1013 hPa)
 :–95 °C
 :1.322 g/cm³ (20 °C)
 :Ambient
 :200-838-9
 :1593
 :6.1,III
 :Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
 :01-2119480404-41
 :GHS08
 :75-09-2
 :CH₂Cl₂
 :MFCD00000881