Order Entry
Austria
ContactUsLinkComponent
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Health
Irritant
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
Certificates
undefined
Health
Irritant
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    20 g/l (20 °C)
  • Melting Point:
    -95 °C
  • Molar Mass:
    84.93 g/mol
  • Boiling Point:
    40 °C (1013 hPa)
  • Vapor Pressure:
    475 hPa (20 °C)
  • Refractive Index:
    1.42
  • Explosion Limit:
    13 - 22 %(V)
  • Density:
    1.33 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    605 °C DIN 51794
  • Synonyms:
    DCM, Methylene chloride, Methylene dichloride
  • Hill Formula:
    CH2Cl2
  • Chemical Formula:
    CH2Cl2
  • LD50 oral:
    LD50 Rat 1600 mg/kg
  • LD50 dermal:
    LD50 Rat greater than 2000 mg/kg
  • Storage:
    Store at +15°C to +25°C

Specification Test Results

Related Information

Frequently Bought Together