Order Entry
Austria
ContactUsLinkComponent
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Health
Irritant
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™
Certificates
undefined
Health
Irritant
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ AMD, Riedel-de Haën™

Specification Test Results

  • Explosion Limit:
    22 %
  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Transmittance at 235 nm:
    Min. 20 %
  • Transmittance at 240 nm:
    Min. 50 %
  • Transmittance at 245 nm:
    Min. 80 %
  • Transmittance from 260 nm:
    Min. 98 %
  • for AMD technic:
    complying
  • Refractive Index:
    n20/D 1.424 (lit.)
  • Autoignition Temperature:
    1223 °F

Specification Test Results

Related Information

Frequently Bought Together