Order Entry
Austria
ContactUsLinkComponent
 
 
 
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
 
undefined
 
 
Dichloromethane ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

 

  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Contains:
    ~25 mg/L amylene as stabilizer
  • Vapor Pressure:
    24.45 psi ( 55 °C)
  • Refractive Index:
    n20/D 1.424 (lit.)
  • Explosion Limit:
    22 %
  • Vapor Density:
    2.9 (vs air)

 

 

Frequently Bought Together