Order Entry
Austria
Orders LinkContactUsLinkComponent
Dichloromethane, BAKER BIO-ANALYZED®, J.T.Baker®

Dichloromethane, BAKER BIO-ANALYZED®, J.T.Baker®

Health
Irritant
Supplier:   Avantor
Dichloromethane, BAKER BIO-ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
Dichloromethane, BAKER BIO-ANALYZED®, J.T.Baker®
Dichloromethane, BAKER BIO-ANALYZED®, J.T.Baker®

2 Options Available Below

Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Assay (by GC):
    min. 99.8 %
  • Color (APHA):
    max. 10
  • Preservative (as Amylene):
    about 20 ppm
  • Residue after Evaporation:
    max. 5 ppm
  • Titrable Acid (meq/g):
    max. 0.0001
  • Water (by KF, coulometric):
    max. 30 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.