

Dichloromethane ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9315-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤233
- Maximum for Impurities, For Information Only:= 99.8 %
- Assay (by GC, corrected for water, exclusive of preservative):≥99.8 %
- Chloride (Cl):≤10 ppm
- Preservative (cyclohexene):≥50 ppm
- Residue after Evaporation:≤2.0 ppm
- Titrable Acid (µeq/g):≤0.3
- Water (by KF, coulometric):≤0.02 %
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
:Packaged under nitrogen.
:Stabilised with cyclohexene ≥ 50 ppm
EINECS:200-838-9
UN:1593
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881