Order Entry
Austria
ContactUsLinkComponent
Dichloromethane ≥99.8% stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Health
Irritant
Dichloromethane ≥99.8% stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Health
Irritant
Dichloromethane ≥99.8% stabilised, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 5
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water):
    ≥99.8 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.3
  • Chloride (Cl):
    ≤10 ppm
  • Water (by KF, coulometric):
    ≤0.02 %

Specification Test Results

Frequently Bought Together