

Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:14,6063
Boiling Pt:39…40 °C
Melting Pt:–95 °C
Density:1.325
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Packaged under argon in resealable ChemSeal™ bottles.
:Stabilised with amylene
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together