

1-Chloropropane for synthesis, Sigma-Aldrich®
: Merck
:540-54-5
:
:C₃H₇Cl
:MFCD00000995
:78.54 g/mol
:12,08032
:46 °C (1013 hPa)
:–122.8 °C
:1.3914 g/cm³ (20 °C)
:208-749-7
:1278
:3,II
:M-Clarity™ quality level = MQ200
:–31 °C
:GHS02
:GHS07
:Refrigerator
- Solubility:2.7 g/l (20 °C)
- Melting Point:-122 °C
- Molar Mass:78.54 g/mol
- Boiling Point:45 - 47 °C
- Vapor Pressure:373 hPa (20 °C)
- Flash Point:-18 °C
- Refractive Index:1.388 (20 °C, 589 nm)
- Explosion Limit:2.6 - 11.1 %(V)
- Density:0.89 g/cm3 (20 °C)
- Ignition Point:520 °C
- Synonyms:Propyl chloride
- Hill Formula:C3H7Cl
Frequently Bought Together