Order Entry
Austria
Orders LinkContactUsLinkComponent
1,1,1-Trichloroethane

1,1,1-Trichloroethane

Catalog #
Irritant
1,1,1-Trichloroethane
Catalog #
Supplier:  
CAS Number:  71-55-6
1,1,1-Trichloroethane
CAS Number:  71-55-6
Product Details & Documents
Synonyms:  1,1,1-TCA, Chlorothene, Methyl chloroform
  • CAS Number:71-55-6
  • Formula:Cl₃CCH₃
  • Molecular Weight:133.4 g/mol
  • Merck Index:12,09766
  • Boiling Pt:74 °C (1013 hPa)
  • Melting Pt:–32.5 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-756-3
  • UN:2831
  • ADR:6.1,III
  •  :Density: 1.31 g/cm³ (20 °C)