Order Entry
Austria
ContactUsLinkComponent
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Irritant
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Certificates
undefined
Irritant
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™

Specification Test Results

  • Form:
    Solid
  • assay:
    68.0 - 69.0 %
  • silver (Ag):
    Max. 10 ppb
  • aluminium (Al):
    Max. 20 ppb
  • gold (Au):
    Max. 10 ppb
  • arsenic (As):
    Max. 5 ppb
  • boron (B):
    Max. 10 ppb
  • barium (Ba):
    Max. 10 ppb
  • beryllium (Be):
    Max. 10 ppb
  • bismuth (Bi):
    Max. 10 ppb
  • calcium (Ca):
    Max. 30 ppb
  • cadmium (Cd):
    Max. 10 ppb
  • cobalt (Co):
    Max. 10 ppb
  • chromium (Cr):
    Max. 30 ppb
  • copper (Cu):
    Max. 10 ppb
  • iron (Fe):
    Max. 100 ppb
  • gallium (Ga):
    Max. 10 ppb
  • germanium (Ge):
    Max. 10 ppb
  • indium (In):
    Max. 10 ppb
  • potassium (K):
    Max. 50 ppb
  • lithium (Li):
    Max. 10 ppb
  • magnesium (Mg):
    Max. 50 ppb
  • manganese (Mn):
    Max. 10 ppb
  • molybdenum (Mo):
    Max. 10 ppb
  • sodium (Na):
    Max. 50 ppb
  • nickel (Ni):
    Max. 30 ppb
  • lead (Pb):
    Max. 10 ppb
  • platinum (Pt):
    Max. 10 ppb
  • antimony (Sb):
    Max. 5 ppb
  • tin (Sn):
    Max. 10 ppb
  • strontium (Sr):
    Max. 10 ppb
  • titanium (Ti):
    Max. 50 ppb
  • thallium (Tl):
    Max. 10 ppb
  • vanadium (V):
    Max. 10 ppb
  • zinc (Zn):
    Max. 10 ppb
  • zirconium (Zr):
    Max. 10 ppb
  • chloride (Cl):
    Max. 0.5 ppm
  • phosphate (PO4):
    Max. 0.5 ppm
  • sulfate (SO4):
    Max. 0.5 ppm

Specification Test Results

Related Information

Frequently Bought Together