Order Entry
Austria
ContactUsLinkComponent
Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC
Irritant
Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153164R
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC
Catalog # 153164R
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 98.0 %
  • pH (20°C; 5 %):
    6.5 - 7.5
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 1 ppm
  • Transmittance (254 nm) (1 mol/l):
    Min. 98.0 %

Specification Test Results

About this item

 :
 :77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
Flash Pt:136 °C
EINECS:211-162-9
 :GHS07
Storage Temperature:Ambient
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066