Potassium acetate 99.0-101.0% Ph. Eur.
:127-08-2
:
:CH₃COOK
:MFCD00012458
:98.14 g/mol
:13,07687
:304 °C
:1.57 g/cm³ (25 °C)
:>250 °C
:Ambient
:204-822-2
- Assay (calculated on dried substance):99.0 - 101.0 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- pH (5 %):7.5 - 9.0
- Reducing substances:Passes test
- Cl (Chloride):Max. 200 ppm
- SO4 (Sulphate):Max. 200 ppm
- Fe (Iron):Max. 20 ppm
- Heavy metals (as Pb):Max. 4 ppm
- Na (Sodium):Max. 0.5 %
- Loss on drying (105°C):Max. 3.0 %
- Residual solvents:Passes test
Frequently Bought Together