Potassium acetate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Specification Test Results
- Assay (calculated on anhydrous):Min. 98 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 0.02 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
26664_POTASSIUM ACETATE GPR RECTAPUR_BSE_REV0
BSE/TSE Certificates
spec_VWRC26664.293 spec_VWRC26664.293 spec_VWRC26664.362 spec_VWRC26664.362
Product specification
Frequently Bought Together