Potassium acetate 99.0-100.5% (dry basis), granular USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Specification Test Results
- GMP Manufactured Product:99.0 - 100.5%
- Meets U.S.P Requirements:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:≤1.0%
- Bulk Pharmaceutical Chemical:7.5 - 8.5
- USP - Assay (CH₃COOK) (dried basis):99.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying:≤1.0 %
- USP - pH (1 in 20):7.5 - 8.5
- USP - Sodium (Na):≤0.03 %
Specification Test Results
Frequently Bought Together