Potassium acetate ≥99% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:14,7605
Melting Pt:292 °C
Density:1.57
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Related Information
Frequently Bought Together