Potassium acetate, anhydrous 99+%, pure
:127-08-2
:
:CH₃COOK
:MFCD00012458
:98.14 g/mol
:13,07687
:292 °C
:1,57 g/cm³ (25 °C)
:>250 °C
:Ambient
:204-822-2
- Appearance:White Crystalline or granular powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥99.0 %
Frequently Bought Together