Potassium acetate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Merck Index:13,07687
Melting Pt:292 °C
Density:1,57 g/cm³ (25 °C)
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
Specification Test Results
- Appearance:White Granular powder
- Infrared spectrum:Conforms
- Water:≤1.0 %
- pH:7.5 to 9.0 (5% soln. at 20°C)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together