Stearic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-11-4
:
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752
:284.48 g/mol
Merck Index:13,08882
Boiling Pt:361 °C (1013 hPa)
Melting Pt:67 °C
Density:0.94 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:196 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-313-4
Frequently Bought Together