Order Entry
Austria
Orders LinkContactUsLinkComponent
Stearic acid

Stearic acid

Marketsource
Supplier:   Roth Carl
Stearic acid
Supplier:  Roth Carl
Marketsource
Certificates
Stearic acid

2 Options Available Below

Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (1013 hPa)
  • Melting Pt:67 °C
  • Density:0.94 g/cm³ (20 °C)
  • Flash Pt:196 °C (closed cup)
  • Storage Temperature:Refrigerator
  • EINECS:200-313-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 34,90 €
    34,90 €
    1 * 1 kg
       34,90 €

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 143,00 €
    143,00 €
    1 * 5 kg
       143,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.