Order Entry
Austria
ContactUsLinkComponent
 
Stearic acid for synthesis, Sigma-Aldrich®
 :  Merck
 
undefined
Stearic acid for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Molar Mass:
    284.49 g/mol
  • Storage:
    Store at +15°C to +25°C
  • Grade:
    REAG
  • Synonyms:
    Octadecanoic Acid
  • Chemical Formula:
    CH3(CH2)16CO2H

 

Frequently Bought Together