Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Stearic acid

Glass bottle for solids, 10 mg
Catalog # CAYM90320-10
Supplier:   Cayman Chemical
Stearic acid
Catalog # CAYM90320-10
Supplier:  Cayman Chemical
CAS Number:  57-11-4
Stearic acid
Stearic acid

Glass bottle for solids, 10 mg

CAS Number:  57-11-4
Price
379,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (1013 hPa)
  • Melting Pt:67 °C
  • Density:0.94 g/cm³ (20 °C)
  • Flash Pt:196 °C (closed cup)
  • Storage Temperature:Refrigerator
  • EINECS:200-313-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications