Stearic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57-11-4
:
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752
:284.48 g/mol
Merck Index:14,8804
Boiling Pt:232 °C (15 mmHg)
Melting Pt:69…72 °C
Density:0.941
Flash Pt:196 °C (384 °F)
EINECS:200-313-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together