Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Formic acid 98-100%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
undefined
 
 
 
Formic acid 98-100%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    4 °C
  • Molar Mass:
    46.03 g/mol
  • Boiling Point:
    101 °C (1013 hPa)
  • Vapor Pressure:
    42 hPa (20 °C)
  • Flash Point:
    48 °C
  • Explosion Limit:
    12 - 38 %(V)
  • Density:
    1.22 g/cm3 (20 °C)
  • pH:
    2.2 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    480 °C

 

Frequently Bought Together