Order Entry
Austria
Orders LinkContactUsLinkComponent
Palmitic acid, Millipore®

Palmitic acid, Millipore®

Plastic bottle for solids, 25 g
Catalog # 506345-25
Palmitic acid, Millipore®
Catalog # 506345-25
CAS Number:  57-10-3
Palmitic acid, Millipore®

Plastic bottle for solids, 25 g

CAS Number:  57-10-3
Price
180,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:13,07064
  • Boiling Pt:352 °C (1013 hPa)
  • Melting Pt:61…63 °C
  • Density:0.853 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-312-9
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.