Heptanoic acid
EHERC14126700
:
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
- Environmentally Preferable:
: Heptylic acid, Enanthic acid
:
:130.19 g/mol
:223
:>110 °C
:203-838-7
:3265
:GHS05
:Ambient
:111-14-8
:C₇H₁₄O₂
:MFCD00004426
:–10 °C
:0.92 g/cm³ (20 °C)
:8 III
:13,04680