Order Entry
Austria
ContactUsLinkComponent
 
Acetic acid 99 - 100% 99-100%, GPR RECTAPUR®
 
undefined
Acetic acid 99 - 100% 99-100%, GPR RECTAPUR®
 

 

  • Environmentally Preferable:

 

 

  • Assay:
    Min. 99.5 %
  • Solidification point:
    16.0 - 16.6 °C
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 50 ppm
  • Non-volatile residue:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm

 

Frequently Bought Together