Order Entry
Austria
Orders LinkContactUsLinkComponent
Acetylacetone 99+%

Acetylacetone 99+%

Toxic
Flammable
Acetylacetone 99+%
Certificates
Acetylacetone 99+%

5 Options Available Below

Product Details & Documents
Synonyms:  ACAC, 2,4-Pentanedione
  • CAS Number:123-54-6
  • Formula:CH₃COCH₂COCH₃
  • MDL Number:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Merck Index:13,00083
  • Boiling Pt:133…135 °C (760 mmHg)
  • Melting Pt:–23 °C
  • Density:0.975 g/cm³
  • Flash Pt:35.5 °C
  • Storage Temperature:Ambient
  • EINECS:204-634-0
  • UN:2310
  • ADR:3,III

Dokumente

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4510 to 1.4530 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤50 APHA
  • Specific gravity:
    (20°C) 0.972 to 0.978
  • Acidity (CH3COOH):
    ≤0.2 %

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,60 €
    29,60 €
    1 * 5 g
       29,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    129960050.
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 37,80 €
    37,80 €
    1 * 250 g
       37,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    129962500.
  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 178,00 €
    178,00 €
    1 * 2,5 kg
       178,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    129960025.
  • Plastic drum
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 581,00 €
    581,00 €
    1 * 10 kg
       581,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    129960100.
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 89,00 €
    89,00 €
    1 * 1 L
       89,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    129960010.