Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650501-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Refractive Index:n20/D 1.359 (lit.)
- Purity (GC):Min. 99.90 %
- GC (as heptachlor epoxide):Max. 10 ng/l
- Water (Karl Fischer):Max. 0.5 %
- Non-volatile matter:Max. 0.0003 %
- IR spectrum:complying
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 375 nm:Max. 0.005
- Absorbance at 350 nm:Max. 0.01
- Absorbance at 340 nm:Max. 0.06
- Absorbance at 330 nm:Max. 1.0
- Fluorescence (chinin) at 365 nm:Max. 1.0 ppb
- Explosion Limit:13.2 %
- Vapor Pressure:184 mmHg ( 20 °C)
Specification Test Results
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:55…57 °C (1.013 hPa)
Melting Pt:–95 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:14.3 %(V)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS07
:GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765