Order Entry
Austria
ContactUsLinkComponent
 
 
 
Acetone ≥99.5% (by GC) for synthesis
 :  Roth Carl
    
 
undefined
 
 
Acetone ≥99.5% (by GC) for synthesis
 :  Roth Carl
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Appearance:
    clear, colourless liquid
  • Assay (GC):
    ≥99,5 %
  • Density (20 °C):
    0,788-0,793 g/cm³
  • Water (KF):
    ≤0,3 %
  • Refractive index (20 °C):
    1,358-1,360

 

Frequently Bought Together