Order Entry
Austria
ContactUsLinkComponent
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Flammable
Irritant
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.2 %
  • Absorbance (345 nm):
    Max. 0.3
  • Absorbance (350 nm):
    Max. 0.1
  • Absorbance (360 nm):
    Max. 0.01
  • Transmittance (345 nm):
    Min. 50 %
  • Transmittance (350 nm):
    Min. 80 %
  • Transmittance (360 nm):
    Min. 98 %

Specification Test Results

Frequently Bought Together