Order Entry
Austria
ContactUsLinkComponent
Acetone, LiChrosolv® for HPLC, Supelco®
Flammable
Irritant
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -95.4 °C
  • Molar Mass:
    58.08 g/mol
  • Boiling Point:
    56.2 °C (1013 hPa)
  • Vapor Pressure:
    233 hPa (20 °C)
  • Refractive Index:
    1.35868 (20 °C)
  • Explosion Limit:
    2.6 - 12.8 %(V)
  • Density:
    0.79 g/cm3 (20 °C)
  • pH:
    5 - 6 (395 g/l, H2O, 20 °C)
  • Ignition Point:
    465 °C DIN 51794
  • Synonyms:
    Dimethyl ketone, Propanone, 2-Propanone
  • Hill Formula:
    C3H6O
  • Chemical Formula:
    CH3COCH3

Specification Test Results

Related Information

Frequently Bought Together