

Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
EINECS:200-662-2
UN:1090
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Purity (GC):=99.8%
- Identity(IR):Conforms
- Evaporation Residue:<= 3.0 mg/L
- Water:= 0.05%
- Colour:=10 Hazen
- GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)):<= 3 pg/ml
- GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)):<= 3 ng/ml
- Suitability:Residue Analysis
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications Merck SupraSolv® High Purity Solvents for Gas Chromatography
Product Literature
msds_100012
SDS
ProductPage_100012
Product specification
Frequently Bought Together