

Acetone, BAKER ANALYZED® HPLC for HPLC, J.T.Baker®
: Avantor
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:01-2119471330-49
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (by GC) (corrected for water):min. 99.7 %
- Aldehyde (as HCHO):max. 0.002 %
- Color (APHA):max. 10
- Isopropyl Alcohol:max. 0.05 %
- Methanol (CH3OH):max. 0.05 %
- Residue after Evaporation:max. 2 ppm
- Solubility in Water:passes test
- Substances Reducing KMnO4:passes test
- Titrable Acid (meq/g):max. 0.0003
- Titrable Base (meq/g):max. 0.0006
- Water (H2O):max. 0.2 %
- at 330 nm:max. 1.00
- at 340 nm:max. 0.10
- at 350 nm:max. 0.01
- at 400 nm:max. 0.01
- UV Cut-off, nm:max. 330
Frequently Bought Together