Order Entry
Austria
ContactUsLinkComponent
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Flammable
Irritant
Health
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Catalog # 5008-05
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Health
Acetone ≥99.5% (by GC), Finyte® for microelectronic, J.T.Baker®
Catalog # 5008-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, pressure bottom
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Microelectronic Use:
    = 99.5 %
  • Assay (by GC):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Acidity (µeq/g):
    ≤0.3
  • Aldehydes (as HCHO):
    ≤20000 ppb
  • Alkalinity (µeq/g):
    ≤0.5
  • Isopropyl Alcohol (CH₃CHOHCH₃)(BY WT):
    ≤0.05 %
  • Methanol (CH₃OH)(BY WT):
    ≤0.05 %
  • Sulfate (SO₄), For Information Only:
    = 3000.0 ppb
  • Residue after Evaporation:
    ≤3000.0 ppb
  • Water (H₂O):
    ≤0.50 %
  • Solubility in H₂O:
    Passes Test
  • Chloride (Cl):
    ≤200.0 ppb
  • Phosphate (PO₄):
    ≤50.0 ppb
  • Trace Impurities - Aluminum (Al):
    ≤10.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤5.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤10.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤10.0 ppb
  • Trace Impurities - Silicon:
    ≤10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤50 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤8 par/ml

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
 :GHS08
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765