Septum caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE with silicone/PTFE septum.
- Sterile 10⁻⁶ SAL
- No adhesives used to attach septum to high density polyethylene closure
- Non-cytotoxic and non-pyrogenic
- Dual septum material for improved gas barrier
Specifications
- Catalog No:
- NALG342178-0384
- NALG342178-0240
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement