Order Entry
Austria
ContactUsLinkComponent
 
Jerricans, flat, with stopcock and screw cap, Lowboys™, Nalgene®
Catalog # NALG2324-0015
   Low Manufacturing Impact
undefined
Jerricans, flat, with stopcock and screw cap, Lowboys™, Nalgene®
Catalog # NALG2324-0015
   Low Manufacturing Impact

Specifications

  • Thread:
    63 mm
  • Capacity:
    15 l
  • Colour:
    Natural
  • W×D×H:
    368×368×203 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Neck I-Ø:
    44 mm
  • Pk:
    1

Specifications

About this item

Made of PP, translucent.

  • Can be stacked three high
  • Autoclave between uses to eliminate bacterial contamination
  • Convenient integral handgrip for safe, easy transport

Carboys have a low-profile design which fits easily into tight spaces, on shelves or in lab refrigerators. Ideal for storing and dispensing reagents.