ContactUsLinkComponent
Acetone for residue analysis
Irritant
Flammable
Acetone for residue analysis
   Sustainable Options Available
Certificates
undefined
Irritant
Flammable
Acetone for residue analysis
   Sustainable Options Available
Certificates
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.2 %
  • GC:
    (ECD) Passes test, (NPD) Passes test, (FID) Passes test
  • Residue after evaporation:
    ≤2 ppm