ContactUsLinkComponent
Vacuum check valve and positive connector, Nalgene®
Vacuum check valve and positive connector, Nalgene®
Catalog # NALG6120-0010
   Low Manufacturing Impact
undefined
Vacuum check valve and positive connector, Nalgene®
Catalog # NALG6120-0010
   Low Manufacturing Impact
Specifications
  • Description:
    Vacuum check valve and positive connector, 19,05 mm length
  • Material:
    HDPE
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Tubing size:
    6,35 - 7,94 mm ID
  • Pk:
    6
Specifications
About this item
These vacuum check valves with positive connector are designed to prevent back pressure with aspirator-type vacuum pumps.
  • Work best between 10- and 28- in. Hg.
  • Silicone diaphragm prevents back pressure