

Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9214-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass, amber, narrow mouth, PVC-coated
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:≤0.07
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 335 nm:≤0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤330
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.5 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (CH₃COCH₂CH₃) (by GC):≥99.5 %
- Residue after Evaporation:≤3.0 ppm
- Water (by KF, volumetric):≤0.03 %
Specification Test Results
About this item
:
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C (closed cup)
ADR:3,II
:Packaged under nitrogen.
:72.11 g/mol
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
REACH:01-2119457290-43
:GHS02
:GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648