ContactUsLinkComponent
Caps for closed dome tanks with mixer support assembly, Nalgene®
Caps for closed dome tanks with mixer support assembly, Nalgene®
Catalog # NALG2651-0200
   Low Manufacturing Impact
undefined
Caps for closed dome tanks with mixer support assembly, Nalgene®
Catalog # NALG2651-0200
   Low Manufacturing Impact
Specifications
  • Cap Size:
    152,3 mm
  • Description:
    with silicone gasket, true union clamp
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    1
  • For bottles:
    75/115/ 210/380 l
  • Environmental certification:
    NAR Certificate of Retirement
Specifications
About this item
Polypropylene, PVDF, silicone.
  • 6" (15,2 cm) polypropylene screw closure with 2" (5,1 cm) sanitary ferrule welded in the center
  • 2" (5,1 cm) silicone gasket and PDVF True Union clamp
  • Autoclavable
Use with Nalgene closed-dome tanks (Series 2650).