Orders LinkContactUsLinkComponent
Buckets
Buckets
Supplier:   Brand
Buckets
Supplier:  Brand
Certificates
Buckets
2 Options Available Below
Product Details & Documents
HDPE, white.
  • Good chemical and temperature resistance
  • Scale with 1 litre steps
  • Without spout and lid (Please order push-on lid seperately)
For industrial and laboratory applications. With handle.
Product Family Options
Product Information
  • Colour
    Capacity
    Height
    Availability
    Price
  • BRND71772
    Bucket, with graduation and handle
    Bucket, with graduation and handle
    White
    5 L
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • BRND71774
    Bucket, with graduation and handle
    Bucket, with graduation and handle
    White
    10 L
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,40 €
    21,40 €
    1 * 1 ST
       21,40 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Push-on lid, LDPE
    Push-on lid, LDPE
    BRND71776
    10 l buckets
    Log in for availability
    1 * 1 ST - 5,90 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Push-on lid, LDPE
    Push-on lid, LDPE
    BRND71771
    5 L buckets
    Log in for availability
    1 * 1 ST - 5,40 €